Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008597.3 | NP_032623.1 | 3, 4 | 250 | ||
GenBank mRNA | AK018719.1 | - | 3, 4 | 251 | ||
AK159591.1 | - | 3, 4 | 254 | |||
AK159931.1 | - | 3, 4 | 251 | |||
BC018338.1 | - | 3, 4 | 142 | |||
BC036991.1 | - | 3, 4 | 246 | 4990627 | ||
BC069922.1 | - | 3, 4 | 240 | 4984262 | ||
BC079478.1 | - | 3, 4 | 241 | 4950210 | ||
BY702829.1 | - | 3, 4 | 251 | |||
D00613.1 | - | 3, 4 | 193 |