Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010806.1 | NP_034936.1 | 19 | 2690 | ||
GenBank mRNA | AK016557.1 | - | 18 | 2809 | ||
CN537179.1 | - | 5 | 560 |