Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010806.1 | NP_034936.1 | 21 | 3108 | ||
GenBank mRNA | CN538841.1 | - | 3 | 329 |