Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008610.2 | NP_032636.1 | 2 | 491 | ||
GenBank mRNA | AK148184.1 | - | 2 | 490 | ||
AK204636.1 | - | 2 | 81 | |||
BC070430.1 | - | 2 | 491 | 6813184 | ||
M84324.1 | - | 2 | 237 |