Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008610.2 | NP_032636.1 | 11 | 2022 | ||
GenBank mRNA | AK148184.1 | - | 11 | 2021 | ||
AK163248.1 | - | 3 | 334 | |||
AK179499.1 | - | 2 | 224 | |||
AK189970.1 | - | 2 | 222 | |||
AK190237.1 | - | 2 | 222 | |||
AK190797.1 | - | 2 | 222 | |||
AK190805.1 | - | 2 | 222 | |||
AK190966.1 | - | 2 | 222 | |||
AK191078.1 | - | 2 | 224 | |||
AK191300.1 | - | 2 | 222 | |||
AK191758.1 | - | 2 | 222 | |||
AK192637.1 | - | 2 | 222 | |||
AK192709.1 | - | 2 | 222 | |||
AK193954.1 | - | 2 | 224 | |||
AK195150.1 | - | 2 | 222 | |||
AK195198.1 | - | 2 | 223 | |||
AK195892.1 | - | 2 | 223 | |||
AK196680.1 | - | 1 | 61 | |||
AK197081.1 | - | 310 | ||||
AK197653.1 | - | 2 | 222 | |||
AK198474.1 | - | 2 | 222 | |||
AK198537.1 | - | 2 | 222 | |||
AK198639.1 | - | 2 | 222 | |||
AK198965.1 | - | 2 | 223 | |||
AK199919.1 | - | 249 | ||||
AK200339.1 | - | 2 | 222 | |||
AK200683.1 | - | 2 | 222 | |||
AK201746.1 | - | 2 | 223 | |||
AK201782.1 | - | 2 | 222 | |||
AK202079.1 | - | 2 | 222 | |||
AK202191.1 | - | 2 | 224 | |||
AK202431.1 | - | 2 | 222 | |||
AK203427.1 | - | 2 | 222 | |||
AK203970.1 | - | 2 | 224 | |||
AK204187.1 | - | 2 | 223 | |||
AK204502.1 | - | 2 | 222 | |||
AK204698.1 | - | 2 | 223 | |||
AK204944.1 | - | 2 | 222 | |||
AK219401.1 | - | 2 | 222 | |||
BC070430.1 | - | 11 | 2022 | 6813184 | ||
M84324.1 | - | 11 | 1768 |