Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008628.2 | NP_032654.1 | 10 | 1689 | ||
GenBank mRNA | AK051960.1 | - | 10 | 1686 | ||
AK157842.1 | - | 10 | 1702 | |||
AK166179.1 | - | 10 | 1677 | |||
BC047117.1 | - | 10 | 1697 | 4951173 | ||
BC050897.1 | - | 10 | 1672 | 6401835 | ||
U21011.1 | - | 10 | 1706 | |||
X81143.1 | - | 10 | 1689 |