Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039368.2 | NP_001034457.1 | 4 | 551 | ||
NM_023127.3 | NP_075616.2 | 4 | 419 | |||
GenBank mRNA | AK008136.1 | - | 4 | 508 | ||
AK008287.1 | - | 4 | 509 | |||
AK018914.1 | - | 4 | 378 |