Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Map6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043355.2 | NP_001036820.2 | 2 | 1051 | ||
NM_001048167.1 | NP_001041632.1 | 2 | 1950 | |||
NM_010837.3 | NP_034967.2 | 2 | 1950 | |||
GenBank mRNA | AK018193.2 | - | 2 | 1430 | ||
AK087413.1 | - | 2 | 1913 | |||
AK154455.1 | - | 2 | 1023 | |||
AK214985.1 | - | 2 | 353 | |||
BC053039.1 | - | 2 | 1951 | 5709455 | ||
BY259340.1 | - | 2 | 273 | |||
Y16032.1 | - | 2 | 991 |