Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161775.1 | NP_001155247.1 | 19 | 2415 | ||
NM_013607.2 | NP_038635.2 | 19 | 2415 | |||
GenBank mRNA | AK137226.1 | - | 20 | 2392 | ||
AK137716.1 | - | 19 | 2371 | |||
AK173045.1 | - | 19 | 2379 | |||
D85923.1 | - | 19 | 2370 | |||
D85924.1 | - | 19 | 2370 |