Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039545.2 | NP_001034634.2 | 3 | 262 | ||
GenBank mRNA | AK036437.1 | - | 3 | 224 | ||
AY963799.1 | - | 3 | 220 | |||
BC008538.1 | - | 2 | 201 | 3495412 | ||
DQ020160.1 | - | 3 | 262 | |||
DQ020161.1 | - | 2 | 319 | |||
DQ020162.1 | - | 3 | 231 |