Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022410.3 | NP_071855.2 | 12 | 1533 | ||
GenBank mRNA | AJ312390.1 | - | 12 | 1457 | ||
AK147203.1 | - | 12 | 1456 | |||
AK147206.1 | - | 12 | 1465 | |||
AK147208.1 | - | 12 | 1473 | |||
AK147209.1 | - | 12 | 1464 | |||
AK147210.1 | - | 12 | 1464 | |||
AK147211.1 | - | 12 | 1508 | |||
AK147215.1 | - | 12 | 1451 | |||
AK147216.1 | - | 12 | 1406 | |||
AK147221.1 | - | 12 | 1411 | |||
AK147222.1 | - | 12 | 1452 | |||
AK147223.1 | - | 12 | 1457 | |||
AK147233.1 | - | 12 | 1464 | |||
AK147235.1 | - | 12 | 1464 | |||
AK147296.1 | - | 12 | 1464 | |||
AK147407.1 | - | 12 | 1467 | |||
AK147430.1 | - | 12 | 1474 |