Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142322.1 | NP_001135794.1 | 26 | 4587 | ||
NM_001142323.1 | NP_001135795.1 | 26 | 4587 | |||
NM_015742.2 | NP_056557.2 | 27 | 4548 | |||
GenBank mRNA | AF143683.1 | - | 27 | 4528 | ||
AK147347.1 | - | 25 | 3777 | |||
BC034526.1 | - | 6 | 823 | |||
BC138454.1 | - | 26 | 4409 | 8861474 |