Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ncoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010881.2 | NP_035011.1 | 20 | 4666 | ||
GenBank mRNA | AK035922.1 | - | 5 | 990 | ||
BC068177.1 | - | 17 | 3974 | 30628581 | ||
U56920.1 | - | 19 | 4224 | |||
U64606.1 | - | 17 | 3969 | |||
U64828.1 | - | 20 | 4579 |