Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HTR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000867.4 | NP_000858.3 | 2 | 573 | ||
NM_001320758.1 | NP_001307687.1 | 2 | 190 | |||
GenBank mRNA | AK313741.1 | - | 2 | 505 | ||
AY136751.1 | - | 1 | 68 | |||
BC063123.1 | - | 2 | 577 | 30321290 | ||
X77307.1 | - | 1 | 123 | |||
Z36748.1 | - | 1 | 114 |
Set Membership: |
![]() |