Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFATC2 SIRNA Assays ›
Gene Symbol:
Nfatc2
Gene Name:
nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2
Gene Aliases:
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp
Chromosome Location:
Chr.2: 168476410 - 168601657 on Build GRCm38
UniGene ID:
Mm.116802
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s70508
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nfatc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037177.2
NP_001032254.1
4
1621
NM_001037178.2
NP_001032255.1
4
986
NM_001136073.2
NP_001129545.1
4
1621
NM_001291168.1
NP_001278097.1
4
1506
NM_001291169.1
NP_001278098.1
4
1506
NM_001291170.1
NP_001278099.1
4
986
NM_001291171.1
NP_001278100.1
3
470
NM_001291172.1
NP_001278101.1
4
1506
NM_001291173.1
NP_001278102.1
3
470
NM_001291174.1
NP_001278103.1
4
986
NM_001291175.1
NP_001278104.1
4
1101
NM_001291176.1
NP_001278105.1
3
585
NM_001291177.1
NP_001278106.1
4
1101
NM_001291178.1
NP_001278107.1
3
585
NM_001291179.1
NP_001278108.1
4
1101
NM_010899.3
NP_035029.2
4
1621
GenBank mRNA
AF289078.1
-
4
1447
AK044489.1
-
4
1507
AK081853.1
-
4
1589
AK083119.1
-
4
962
AK161174.1
-
2
196
BC064006.1
-
4
1540
30290121
EU887581.1
-
4
1387
EU887582.1
-
4
1447
EU887583.1
-
4
984
EU887584.1
-
4
1084
EU887585.1
-
3
468
EU887586.1
-
3
568
EU887587.1
-
4
1387
EU887588.1
-
4
1447
EU887589.1
-
4
984
EU887590.1
-
4
1084
EU887591.1
-
3
468
EU887592.1
-
3
568
EU887599.1
-
4
1387
EU887600.1
-
4
1447
EU887601.1
-
4
984
EU887602.1
-
4
1084
EU887603.1
-
3
468
EU887604.1
-
3
568
U36575.1
-
4
1517
U36576.1
-
4
1517
Back To Top
More Information
Panther Classification:
Molecular Function -
Rel homology transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
positive regulation of gene expression
myotube cell development
cell migration
positive regulation of B cell proliferation
calcineurin-NFAT signaling cascade
response to drug
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
B cell receptor signaling pathway
positive regulation of myoblast fusion
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0