Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081981.2 | NP_001075450.1 | 5 | 1753 | ||
NM_001081982.2 | NP_001075451.1 | 5 | 1078 | |||
NM_001297601.1 | NP_001284530.1 | 5 | 1753 | |||
NM_010906.3 | NP_035036.1 | 5 | 1078 | |||
GenBank mRNA | AK158568.1 | - | 5 | 1756 | ||
BC003766.1 | - | 5 | 800 | 3491917 | ||
BC090626.1 | - | 5 | 780 | |||
S81451.1 | - | 4 | 764 | |||
U57636.1 | - | 4 | 755 | |||
Y07688.1 | - | 5 | 1080 | |||
Y07689.1 | - | 5 | 882 |