Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004507.3 | NP_004498.1 | 6, 7 | 748 | ||
NR_037917.1 | 6, 7 | 806 | ||||
XM_017012115.1 | XP_016867604.1 | 6, 7 | 746 | |||
GenBank mRNA | AF076844.1 | - | 6, 7 | 705 | ||
AF110393.1 | - | 6, 7 | 702 | |||
AJ227901.1 | - | 2, 3 | 163 | |||
AK290459.1 | - | 6, 7 | 685 | |||
AK294117.1 | - | 6, 7 | 734 | |||
BC007013.1 | - | 6, 7 | 666 | 3684398 | ||
BT019481.1 | - | 6, 7 | 642 | |||
BT019482.1 | - | 6, 7 | 642 | |||
CR536552.1 | - | 6, 7 | 642 | |||
Y16893.1 | - | 6, 7 | 642 |