Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008710.3 | NP_032736.2 | 2944 | |||
GenBank mRNA | AK033282.1 | - | 13 | 2132 | ||
AK040460.1 | - | 11 | 1810 | |||
AK050478.1 | - | 12 | 2849 | |||
AK052234.1 | - | 13 | 2176 | |||
AK054332.1 | - | 13 | 2177 | |||
AK083413.1 | - | 13 | 2175 | |||
AK084546.1 | - | 13 | 2174 | |||
AK084604.1 | - | 13 | 2174 | |||
AK085903.1 | - | 11 | 1835 | |||
AK088044.1 | - | 13 | 2926 | |||
AK090115.1 | - | 2947 | ||||
AK129006.1 | - | 13 | 2921 | |||
AK159694.1 | - | 15 | 2930 | |||
AK168736.1 | - | 12 | 2727 | |||
BC008518.1 | - | 13 | 2907 | 3601702 | ||
Z49204.1 | - | 12 | 2851 |