Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008731.3 | NP_032757.2 | 2 | 485 | ||
GenBank mRNA | AK018543.1 | - | 2 | 462 | ||
AK049892.1 | - | 2 | 473 | |||
BC104490.1 | - | 2 | 379 | 40047004 | ||
BC104734.1 | - | 2 | 360 | 40047007 | ||
D86238.1 | - | 2 | 496 |
Set Membership: |
![]() |