Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Npy2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205099.1 | NP_001192028.1 | 3 | 616 | ||
NM_008731.3 | NP_032757.2 | 2 | 955 | |||
GenBank mRNA | AK018543.1 | - | 2 | 932 | ||
AK049892.1 | - | 2 | 943 | |||
BC104490.1 | - | 2 | 849 | 40047004 | ||
BC104733.1 | - | 4 | 491 | 40047010 | ||
BC104734.1 | - | 2 | 830 | 40047007 | ||
D86238.1 | - | 2 | 966 |
Set Membership: |
![]() |