Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146161.1 | NP_001139633.1 | 15 | 1612 | ||
NM_008732.2 | NP_032758.2 | 15 | 1612 | |||
GenBank mRNA | AF029758.1 | - | 15 | 1547 | ||
AK049856.1 | - | 15 | 1560 | |||
AK083478.1 | - | 15 | 1558 | |||
AK148276.1 | - | 15 | 1560 | |||
BC019137.1 | - | 15 | 1586 | 5040976 | ||
L33415.1 | - | 15 | 1540 |