Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146161.1 | NP_001139633.1 | 7 | 703 | ||
NM_008732.2 | NP_032758.2 | 7 | 703 | |||
GenBank mRNA | AF029758.1 | - | 7 | 638 | ||
AK049856.1 | - | 7 | 651 | |||
AK083478.1 | - | 7 | 649 | |||
AK148276.1 | - | 7 | 651 | |||
AK213433.1 | - | 3 | 185 | |||
BC019137.1 | - | 7 | 677 | 5040976 | ||
L33415.1 | - | 7 | 631 |