Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190187.1 | NP_001177116.1 | 5, 6 | 1457 | ||
NM_001190188.1 | NP_001177117.1 | 5, 6 | 1460 | |||
NM_008734.3 | NP_032760.1 | 5, 6 | 1460 | |||
GenBank mRNA | AF010130.1 | - | 5, 6 | 1460 | ||
BC137838.1 | - | 5, 6 | 1191 | 8860857 | ||
BC144964.1 | - | 5, 6 | 1197 | 9053512 |