Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020252.3 | NP_064648.3 | 14 | 3909 | ||
NM_177284.2 | NP_796258.2 | 12 | 3873 | |||
GenBank mRNA | AB093249.2 | - | 14 | 3909 | ||
AK133271.1 | - | 9 | 2500 | |||
AK137407.1 | - | 13 | 3893 | |||
BC079893.1 | - | 12 | 3804 |