Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018766.2 | NP_061236.1 | 2 | 1205 | ||
GenBank mRNA | AB017027.1 | - | 2 | 1206 | ||
BC087730.1 | - | 2 | 1132 | |||
BC138630.1 | - | 2 | 954 | 8861650 | ||
BC138633.1 | - | 2 | 954 | 8861653 |
Set Membership: |
G-protein coupled receptor |