Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053074.1 | NP_444304.1 | 2 | 1034 | ||
GenBank mRNA | AK147894.1 | - | 2 | 1063 | ||
AK149637.1 | - | 2 | 1085 | |||
AK150087.1 | - | 2 | 1055 | |||
AK155059.1 | - | 2 | 1059 | |||
AK166621.1 | - | 2 | 1202 | |||
AK168166.1 | - | 2 | 1056 | |||
AK168486.1 | - | 2 | 1067 | |||
AK169178.1 | - | 2 | 1078 | |||
AK171455.1 | - | 2 | 1369 | |||
AK171611.1 | - | 2 | 1367 | |||
BC005784.1 | - | 2 | 1034 | 3603287 | ||
BC089317.1 | - | 2 | 1115 | 6829463 | ||
S59342.1 | - | 2 | 1032 |