Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301034.1 | NP_001287963.1 | 1 | 210 | ||
NM_008753.4 | NP_032779.2 | 1 | 210 | |||
GenBank mRNA | AA270439.1 | - | 1 | 183 | ||
AK003452.1 | - | 1 | 182 | |||
AK010615.1 | - | 1 | 181 | |||
AK019155.1 | - | 1 | 181 | |||
AK087942.1 | - | 1 | 186 | |||
AK146571.1 | - | 1 | 181 | |||
AK147167.1 | - | 1 | 181 | |||
AK151635.1 | - | 1 | 181 | |||
AK159849.1 | - | 1 | 181 | |||
AK159874.1 | - | 1 | 181 | |||
AK160491.1 | - | 1 | 182 | |||
AK160598.1 | - | 1 | 181 | |||
AK169201.1 | - | 1 | 181 | |||
BC002132.1 | - | 1 | 146 | |||
BC024902.1 | - | 1 | 183 | |||
BC046290.1 | - | 1 | 173 | |||
BC060035.1 | - | 1 | 148 | |||
BC091773.1 | - | 1 | 143 | |||
BC094287.1 | - | 1 | 169 | 5257966 | ||
BY112600.1 | - | 1 | 212 | |||
U52822.1 | - | 1 | 112 |