Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008756.2 | NP_032782.1 | 3 | 315 | ||
GenBank mRNA | AK019880.1 | - | 3 | 267 | ||
AK033532.1 | - | 3 | 248 | |||
AK042576.1 | - | 3 | 315 | |||
BC138679.1 | - | 3 | 298 | 8861700 | ||
BC138680.1 | - | 3 | 298 | 8861701 | ||
BC145371.1 | - | 3 | 313 | 9053929 | ||
U49185.1 | - | 3 | 358 |