Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320605.1 | NP_001307534.1 | 5 | 1168 | ||
NM_001320606.1 | NP_001307535.1 | 6 | 1753 | |||
NM_001320608.1 | NP_001307537.1 | 5 | 1773 | |||
NM_002162.4 | NP_002153.2 | 6 | 1456 | |||
GenBank mRNA | AF340060.1 | - | 6 | 1209 | ||
AF340061.1 | - | 6 | 1209 | |||
AK226023.1 | - | 6 | 1290 | |||
AK309643.1 | - | 6 | 1538 | |||
AK312826.1 | - | 6 | 1291 | |||
AX746321.1 | - | 6 | 1251 | |||
BC040123.1 | - | 6 | 1236 | |||
BC046121.1 | - | 3 | 2002 | |||
BC058903.1 | - | 6 | 1242 | 5207143 | ||
S50015.1 | - | 6 | 1237 | |||
X69711.1 | - | 6 | 1251 | |||
X69819.1 | - | 6 | 1244 |