Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111073.2 | NP_001104543.1 | 2 | 473 | ||
NM_001287213.1 | NP_001274142.1 | 3 | 345 | |||
NM_001287217.1 | NP_001274146.1 | 3 | 333 | |||
NM_008761.4 | NP_032787.1 | 3 | 333 | |||
GenBank mRNA | AI121044.1 | - | 3 | 333 | ||
AK003068.1 | - | 3 | 116 | |||
AK155763.1 | - | 2 | 453 | |||
AK169966.1 | - | 3 | 116 | |||
AK170810.1 | - | 3 | 121 | |||
BC013340.1 | - | 3 | 119 | |||
BC031112.1 | - | 3 | 102 | |||
CA528582.1 | - | 3 | 324 | |||
U72680.1 | - | 3 | 139 |