Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146870.2 | NP_667081.2 | 1 | 770 | ||
GenBank mRNA | BC125306.1 | - | 1 | 770 | 40129821 | |
BC125308.1 | - | 1 | 770 | 40129823 |