Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001545.2 | NP_001536.1 | 5, 6 | 557 | ||
XM_017024574.1 | XP_016880063.1 | 5, 6 | 550 | |||
GenBank mRNA | AK222553.1 | - | 5, 6 | 547 | ||
AK314138.1 | - | 5, 6 | 545 | |||
BC015335.1 | - | 5, 6 | 540 | 4418983 | ||
BT007111.1 | - | 5, 6 | 534 | |||
X81788.1 | - | 5, 6 | 536 |