Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146315.2 | NP_666427.2 | 1 | 281 | ||
GenBank mRNA | AF102541.1 | - | 1 | 83 | ||
BC104294.1 | - | 1 | 280 | |||
BC104295.1 | - | 1 | 282 | 40042931 |