Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Omp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011010.2 | NP_035140.1 | 1 | 237 | ||
GenBank mRNA | AK133724.1 | - | 1 | 268 | ||
BC046600.1 | - | 1 | 237 | 6312008 | ||
BC058228.1 | - | 1 | 257 | |||
BC132327.1 | - | 1 | 236 | 40130604 | ||
BC138222.1 | - | 1 | 236 | 8861241 |