Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008774.3 | NP_032800.2 | 3 | 958 | ||
GenBank mRNA | AK005009.1 | - | 3 | 959 | ||
AK044829.1 | - | 3 | 955 | |||
AK089747.1 | - | 3 | 958 | |||
AK133196.1 | - | 3 | 959 | |||
AK150839.1 | - | 3 | 959 | |||
AK152064.1 | - | 3 | 926 | |||
AK159703.1 | - | 3 | 958 | |||
AK159733.1 | - | 3 | 958 | |||
AK160883.1 | - | 3 | 958 | |||
AK160968.1 | - | 3 | 957 | |||
AK161123.1 | - | 3 | 961 | |||
AK161671.1 | - | 3 | 957 | |||
AK168466.1 | - | 3 | 957 | |||
BC003870.1 | - | 3 | 577 | 3491915 | ||
BC004587.1 | - | 2 | 164 | |||
BC011207.1 | - | 3 | 715 | 4022169 | ||
BC023145.1 | - | 3 | 553 | 3988101 | ||
BC046233.1 | - | 3 | 948 | 6816124 | ||
X65553.1 | - | 3 | 744 |