Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013626.3 | NP_038654.2 | 11 | 1199 | ||
GenBank mRNA | AK051649.1 | - | 11 | 1189 | ||
U79523.1 | - | 11 | 1084 |