Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011041.2 | NP_035171.1 | 2 | 698 | ||
GenBank mRNA | AK034985.1 | - | 2 | 695 | ||
AK132404.1 | - | 2 | 699 | |||
BC005794.1 | - | 2 | 638 | 3707718 | ||
BC006930.1 | - | 1 | 1320 | |||
X84000.1 | - | 2 | 670 |