Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042726.3 | NP_001036191.1 | 2, 3 | 2741 | ||
NM_021543.4 | NP_067518.2 | 2, 3 | 3032 | |||
GenBank mRNA | AF231125.1 | - | 2, 3 | 2956 | ||
AK048638.1 | - | 2, 3 | 2742 | |||
AK140407.1 | - | 2, 3 | 2629 | |||
BC053008.1 | - | 2, 3 | 3020 | 5698009 |