Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162946.1 | NP_001156418.1 | 13 | 1690 | ||
NM_008797.3 | NP_032823.2 | 12 | 1667 | |||
GenBank mRNA | AK077665.1 | - | 4 | 495 | ||
AK128927.1 | - | 1617 | ||||
AK148328.1 | - | 13 | 1637 | |||
AK170596.1 | - | 13 | 1650 | |||
AK170871.1 | - | 13 | 1640 | |||
AK172320.1 | - | 13 | 1618 | |||
AK182295.1 | - | 2 | 58 | |||
BC055030.1 | - | 13 | 1625 | 5053475 | ||
L09192.1 | - | 12 | 1643 | |||
M97957.1 | - | 7 | 815 |