Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011051.3 | NP_035181.1 | 4 | 439 | ||
GenBank mRNA | AK008610.1 | - | 4 | 363 | ||
AK028223.1 | - | 4 | 394 | |||
AK078044.1 | - | 4 | 367 | |||
AK147979.1 | - | 4 | 397 | |||
AK150335.1 | - | 4 | 357 | |||
BC040079.1 | - | 4 | 390 | 3156800 | ||
BU604816.1 | - | 4 | 442 | |||
U49112.1 | - | 4 | 376 | |||
X14938.1 | - | 3 | 206 |