Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162415.1 | NP_001155887.1 | 12 | 1301 | ||
NM_001162416.1 | NP_001155888.1 | 10 | 1094 | |||
NM_008825.4 | NP_032851.2 | 11 | 1195 | |||
GenBank mRNA | AK029659.1 | - | 12 | 1302 | ||
AK146744.1 | - | 11 | 1189 | |||
BC018418.1 | - | 11 | 1152 | 3979400 | ||
BC051014.1 | - | 10 | 1108 | 6493944 | ||
EU616666.1 | - | 9 | 968 | |||
EU616667.1 | - | 10 | 1093 | |||
EU616668.1 | - | 10 | 1093 | |||
EU616669.1 | - | 10 | 1093 | |||
EU616670.1 | - | 9 | 959 | |||
X98847.1 | - | 10 | 1113 |