Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011076.2 | NP_035206.2 | 10 | 1143 | ||
GenBank mRNA | AY864315.1 | - | 9 | 1006 | ||
M24417.1 | - | 4 | 492 | |||
M30697.1 | - | 10 | 1157 | |||
M33581.1 | - | 10 | 1142 |