Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024955.2 | NP_001020126.1 | 7 | 1139 | ||
NM_001077495.2 | NP_001070963.1 | 13 | 2228 | |||
GenBank mRNA | AK046259.1 | - | 7 | 1142 | ||
BC026146.1 | - | 13 | 2209 | 3979333 | ||
BC051106.1 | - | 7 | 839 | 5031261 | ||
M60651.1 | - | 13 | 2163 | |||
U50413.1 | - | 13 | 2181 |