Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008841.2 | NP_032867.2 | 9, 10 | 1580 | ||
GenBank mRNA | AK146195.1 | - | 9, 10 | 1570 | ||
AK148122.1 | - | 9, 10 | 1577 | |||
BC006796.1 | - | 9, 10 | 1571 | 2581969 | ||
BC085501.1 | - | 8, 9 | 1503 | 6816009 | ||
Y13569.1 | - | 9, 10 | 1549 |