Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002169.2 | NP_002160.1 | 1 | 167 | ||
GenBank mRNA | BC093755.1 | - | 1 | 167 | 7939600 | |
BC093757.1 | - | 1 | 167 | 7939602 |