Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146687.2 | NP_001140159.1 | 7 | 971 | ||
NM_001293646.1 | NP_001280575.1 | 7 | 971 | |||
NM_001293647.1 | NP_001280576.1 | 7 | 984 | |||
NM_008844.3 | NP_032870.2 | 7 | 971 | |||
GenBank mRNA | AB006916.1 | - | 7 | 954 | ||
AK014403.1 | - | 7 | 937 | |||
AK041402.1 | - | 7 | 957 | |||
AK122319.2 | - | 7 | 929 | |||
AK154816.1 | - | 7 | 963 | |||
AK155371.1 | - | 7 | 947 | |||
AK170286.1 | - | 7 | 963 | |||
AK171576.1 | - | 7 | 945 | |||
AK185426.1 | - | 1 | 104 | |||
BC019138.1 | - | 7 | 920 | 5041431 | ||
BC020365.1 | - | 7 | 933 | |||
BC094665.1 | - | 7 | 943 | 30356181 |