Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164198.1 | NP_001157670.1 | 10 | 1245 | ||
NM_001164199.1 | NP_001157671.1 | 10 | 1118 | |||
NM_001164200.1 | NP_001157672.1 | 10 | 1210 | |||
NM_011100.4 | NP_035230.1 | 10 | 1249 | |||
GenBank mRNA | AK035560.1 | - | 3 | 333 | ||
AK048319.1 | - | 10 | 1241 | |||
AK081984.1 | - | 3 | 259 | |||
AK139419.1 | - | 10 | 990 | |||
AK148728.1 | - | 10 | 1059 | |||
AK150122.1 | - | 10 | 1211 | |||
AK150134.1 | - | 10 | 1211 | |||
AK163585.1 | - | 10 | 1023 | |||
BC054533.1 | - | 10 | 1233 | 5704796 | ||
J02626.1 | - | 10 | 1178 | |||
X61434.1 | - | 2 | 148 |