Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008855.2 | NP_032881.1 | 10 | 1321 | ||
GenBank mRNA | AK051862.1 | - | 10 | 1314 | ||
BC127083.1 | - | 10 | 1140 | 40111744 | ||
X53532.1 | - | 10 | 1149 | |||
X59274.1 | - | 10 | 1137 |