Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IFNA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002170.3 | NP_002161.2 | 1 | 81 | ||
GenBank mRNA | BC104828.1 | - | 1 | 80 | 8143831 | |
BC104830.1 | - | 1 | 80 | 8143833 | ||
K01900.1 | - | 1 | 95 | |||
V00545.1 | - | 1 | 82 | |||
V00550.1 | - | 1 | 82 |