Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000874.4 | NP_000865.2 | 7 | 980 | ||
NM_001289125.1 | NP_001276054.1 | 7 | 980 | |||
NM_001289126.1 | NP_001276055.1 | 7 | 934 | |||
NM_001289128.1 | NP_001276057.1 | 7 | 980 | |||
NM_207584.2 | NP_997467.1 | 7 | 934 | |||
NM_207585.2 | NP_997468.1 | 7 | 934 | |||
GenBank mRNA | AK293059.1 | - | 6 | 617 | ||
BC002793.2 | - | 7 | 928 | 3626374 | ||
CR541817.1 | - | 6 | 575 | |||
HG380509.1 | - | 7 | 700 | |||
L41942.1 | - | 7 | 901 | |||
L41943.1 | - | 6 | 614 | |||
L41944.1 | - | 7 | 901 | |||
U29584.1 | - | 7 | 641 | |||
X77722.1 | - | 7 | 800 | |||
X89772.1 | - | 6 | 575 | |||
X89814.1 | - | 7 | 739 |